Order Entry
Finland
ContactUsLinkComponent
3-Methyl-1,2-butadiene ≥97.0% (by GC)
 
 
 
3-Methyl-1,2-butadiene ≥97.0% (by GC)
  TCIAM0621-1ML
 :  TCI
 :  
undefined
 
 
 
3-Methyl-1,2-butadiene ≥97.0% (by GC)
  TCIAM0621-1ML
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • Seq:
    2070

 

 

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 97.0 %

 

 

 :
 :68.12 g/mol
 :40°C
 :–113.6 °C
 :0,6831 g/cm³ (25 °C)
 :-12°C
 :–12°C
 :Refrigerator
 :209-926-1
 :3295
 :3,II
 :GHS02
 :GHS07
 :GHS08
 :598-25-4
 :C₅H₈
 :MFCD00008896