
2-Pyrrolecarbaldehyde ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:1003-29-8
:
Formula:C₅H₅NO
MDL Number:MFCD00005217
:95.1 g/mol
Boiling Pt:219°C
Melting Pt:43…46 °C
EINECS:213-705-5
:GHS07
Specification Test Results
- Appearance:White to Brown powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:44.0 to 48.0 deg-C