Order Entry
Finland
ContactUsLinkComponent
 
1,1,1-Trichloro-2-methyl-2-propanol hemihydrate ≥98.0% (by GC, titration analysis)
 :  TCI
 
undefined
1,1,1-Trichloro-2-methyl-2-propanol hemihydrate ≥98.0% (by GC, titration analysis)
 :  TCI

 

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (Argentmetric Titration):
    min. 98.0 % (calcd.on anh.substance)
  • Solubility in EtOH:
    almost transparency
  • Water:
    4.0 to 6.0 %