Order Entry
Finland
ContactUsLinkComponent
2-Amino-6-bromobenzothiazole ≥97.0% (by GC, titration analysis)
Irritant
2-Amino-6-bromobenzothiazole ≥97.0% (by GC, titration analysis)
Catalog # TCIAA2379-5G
Supplier:  TCI
CAS Number:  
undefined
Irritant
2-Amino-6-bromobenzothiazole ≥97.0% (by GC, titration analysis)
Catalog # TCIAA2379-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    White to Light yellow to Light orange powder to crystal
  • Purity (GC):
    min. 97.0 %
  • Purity (Nonaqueous Titration):
    min. 97.0 %

Specification Test Results

About this item

 :
 :229.1 g/mol
Melting Pt:214°C
 :GHS07
CAS Number:15864-32-1
Formula:C₇H₅BrN₂S
MDL Number:MFCD00152229