Order Entry
Finland
ContactUsLinkComponent
2-Methyl-1-butanethiol ≥95.0% (by GC)
Flammable
Irritant
2-Methyl-1-butanethiol ≥95.0% (by GC)
Catalog # TCIAM1041-5G
Supplier:  TCI
CAS Number:  
undefined
Flammable
Irritant
2-Methyl-1-butanethiol ≥95.0% (by GC)
Catalog # TCIAM1041-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 95.0 %

Specification Test Results

About this item

 :
 :104.22 g/mol
Boiling Pt:119°C
Density:0,8483 g/cm³ (13 °C)
Flash Pt:19°C
EINECS:217-515-3
UN:3336
ADR:3,II
 :GHS02
 :GHS07
CAS Number:1878-18-8
Formula:C₅H₁₂S
MDL Number:MFCD00004883