Order Entry
Finland
ContactUsLinkComponent
 
 
2-(4-Fluorophenyl)ethanol ≥97.0% (by GC)
  TCIAF0624-5G
 :  TCI
 :  
undefined
 
2-(4-Fluorophenyl)ethanol ≥97.0% (by GC)
  TCIAF0624-5G
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

 

 

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (GC):
    min. 97.0 %

 

 

 :
 :140.16 g/mol
 :118°C
 :1,137 g/cm³ (20 °C)
 :86°C
 :231-499-5
 :GHS07
 :7589-27-7
 :C₈H₉FO
 :MFCD00002898