
2-(3-Fluorophenyl)ethanol ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:52059-53-7
:
Formula:C₈H₉FO
MDL Number:MFCD00045998
:140.16 g/mol
Boiling Pt:90°C
Density:1,12 g/cm³ (20 °C)
Flash Pt:107°C
:GHS07
Specification Test Results
- Appearance:Light yellow to Yellow to Orange clear liquid
- Purity (GC):min. 98.0 %