Order Entry
Finland
ContactUsLinkComponent
 
 
 
6-Methoxy-m-toluidine ≥98.0% (by GC, titration analysis)
 :  TCI
 
undefined
 
 
6-Methoxy-m-toluidine ≥98.0% (by GC, titration analysis)
 :  TCI

 

  • Appearance:
    White to Amber to Dark green powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Melting point:
    51.0 to 54.0 deg-C