
(Methyl)triphenylphosphonium iodide ≥98.0% (by titrimetric analysis)
: TCI
:2065-66-9
:
:C₁₉H₁₈IP
:MFCD00066175
:404.23 g/mol
:193…194 °C
:218-178-5
:6.1,III
:GHS07
- Appearance:White to Almost white powder to crystal
- Purity (Argentmetric Titration):min. 98.0 %
- Melting point:185.0 to 189.0 deg-C
- Solubility in Methanol:almost transparency
Frequently Bought Together