
(±)-4-Chloromandelic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:492-86-4
:
Formula:C₈H₇ClO₃
MDL Number:MFCD00042724
:186.59 g/mol
Boiling Pt:361 °C (1013 hPa)
Melting Pt:103…105 °C
EINECS:207-764-6
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:118.0 to 122.0 deg-C