G418 disulphate ≥90.0% (by HPLC, total nitrogen)
Supplier: TCI
Certificates
About this item
CAS Number:108321-42-2
:
Formula:C₂₀H₄₄N₄O₁₈S₂
MDL Number:MFCD00058314
:692.72 g/mol
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 90.0 area%
- Purity (with Total Nitrogen):min. 90.0 %
- Specific rotation [a]20/D:+110 to +120 deg (C=0.3, H2O, calcd.on anh.substance)