Order Entry
Finland
ContactUsLinkComponent
2-Methoxypyrazine ≥98.0% (by GC)
Flammable
Irritant
2-Methoxypyrazine ≥98.0% (by GC)
Catalog # TCIAM0798-5ML
Supplier:  TCI
CAS Number:  
undefined
Flammable
Irritant
2-Methoxypyrazine ≥98.0% (by GC)
Catalog # TCIAM0798-5ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 mL
  • Seq:
    2210

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 %

Specification Test Results

About this item

 :
 :110.12 g/mol
Boiling Pt:68°C
Density:1,143 g/cm³ (20 °C)
Flash Pt:50°C
EINECS:221-579-8
UN:1993
ADR:3,III
 :GHS02
 :GHS07
CAS Number:3149-28-8
Formula:C₅H₆N₂O
MDL Number:MFCD00006126