
1-Hydroxy-2-naphthoic acid ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:86-48-6
:
Formula:C₁₁H₈O₃
MDL Number:MFCD00003960
:188.18 g/mol
Merck Index:14,04834
Boiling Pt:375 °C (1013 hPa)
Melting Pt:205 °C
EINECS:201-674-0
ADR:6.1,III
:GHS07
Specification Test Results
- Appearance:White to Light red to Green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Solubility in Methanol:very faint turbidity