
4-Chloro-2-fluorophenol ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:348-62-9
:
Formula:C₆H₄ClFO
MDL Number:MFCD00075030
:146.55 g/mol
Boiling Pt:104°C
Melting Pt:20°C
Flash Pt:81°C
:GHS07
Specification Test Results
- Appearance:White or Colorless to Light yellow powder to lump to clear liquid
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
Specification Test Results
Frequently Bought Together