
2-Bromo-4-chloro-6-fluoroaniline ≥98.0% (by GC)
Catalog # TCIAB2723-5G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:Purple to Dark purple to Dark red powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:40.0 to 44.0 deg-C
Specification Test Results
About this item
:
:224.46 g/mol
:GHS07
CAS Number:195191-47-0
Formula:C₆H₄BrClFN
MDL Number:MFCD00070746