
3-Acetylthiophene ≥95.0% (by GC)
TCIAA1006-10G
: TCI
:
- Pack type:Glass bottle
- Pk:10 g
- Seq:3236
- Appearance:White to Light yellow to Dark green powder to crystal
- Purity (GC):min. 95.0 %
- Melting point:59.0 to 62.0 deg-C
- Solubility in Methanol:very faint turbidity
:
:126.18 g/mol
:95°C
:56 °C
:215-996-4
:6.1,III
:GHS07
:1468-83-3
:C₆H₆OS
:MFCD00005468