
2-Naphthoic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:93-09-4
:
Formula:C₁₁H₈O₂
MDL Number:MFCD00004101
:172.18 g/mol
Merck Index:14,06382
Boiling Pt:333 °C (1013 hPa)
Melting Pt:180…183 °C
Density:1,382 g/cm³ (20 °C)
EINECS:202-217-8
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:184.0 to 187.0 deg-C
- Solubility in Methanol:almost transparency