Order Entry
Finland
ContactUsLinkComponent
 
4,4-Dimethyl-2-cyclohexen-1-one ≥96.0% (by GC)
 :  TCI
 
undefined
4,4-Dimethyl-2-cyclohexen-1-one ≥96.0% (by GC)
 :  TCI

 

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (GC):
    min. 96.0 %