
5-(Hydroxymethyl)resorcinol ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:29654-55-5
:
Formula:C₇H₈O₃
MDL Number:MFCD00016867
:140.14 g/mol
EINECS:249-751-8
:GHS07
Specification Test Results
- Appearance:White to Light yellow to Light red powder to crystal
- Purity (GC):min. 97.0 %
- Melting point:184.0 to 187.0 deg-C
- Solubility in Methanol:very faint turbidity