
2-Nitro-α,α,α-trifluoro-p-toluic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:320-94-5
:
Formula:C₈H₄F₃NO₄
MDL Number:MFCD00007142
:235.12 g/mol
:GHS07
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:135.0 to 139.0 deg-C