α-Bromo-3-chloro-5-fluorotoluene ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:493024-39-8
:
Formula:C₇H₅BrClF
MDL Number:MFCD03788472
:223.47 g/mol
UN:3265
ADR:8,II
:GHS05
Specification Test Results
- Appearance:Colorless to Light orange to Yellow clear liquid
- Purity (GC):min. 98.0 %