
α-Bromo-2-chloro-6-fluorotoluene ≥98.0% (by GC)
: TCI
:68220-26-8
:
:C₇H₅BrClF
:MFCD00040126
:223.47 g/mol
:223 °C (1013 hPa)
:37 °C
:1,629 g/cm³ (20 °C)
:3265
:8,II
:GHS05
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 98.0 %