
ω-Bromo-4-chloroacetophenone ≥98.0% (by GC, titration analysis)
: TCI
:536-38-9
:
:C₈H₆BrClO
:MFCD00000625
:233.49 g/mol
:14,02153
:278 °C (1013 hPa)
:110 °C
:208-631-5
:3261
:8,III
:GHS05
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Argentmetric Titration):min. 98.0 %
- Melting point:96.0 to 100.0 deg-C
- Solubility in Methanol:almost transparency
Frequently Bought Together