Order Entry
Finland
ContactUsLinkComponent
 
 
Methyl-4-iodobenzoate ≥98.0% (by GC)
 :  TCI
 
undefined
 
Methyl-4-iodobenzoate ≥98.0% (by GC)
 :  TCI

 

  • Appearance:
    White to Light yellow to Light red powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    113.0 to 117.0 deg-C
  • Solubility in hot Methanol:
    almost transparency

 

Frequently Bought Together