Order Entry
Finland
ContactUsLinkComponent
 
4-Cyanobenzenesulphonyl chloride ≥98.0% (by GC, titration analysis)
 :  TCI
 
undefined
4-Cyanobenzenesulphonyl chloride ≥98.0% (by GC, titration analysis)
 :  TCI

 

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (Argentmetric Titration):
    min. 98.0 %
  • Melting point:
    108.0 to 112.0 deg-C