Order Entry
Finland
ContactUsLinkComponent
cis-2-Hexen-1-ol ≥93.0% (by GC)
Flammable
cis-2-Hexen-1-ol ≥93.0% (by GC)
Catalog # TCIAH0751-25ML
Supplier:  TCI
CAS Number:  
undefined
Flammable
cis-2-Hexen-1-ol ≥93.0% (by GC)
Catalog # TCIAH0751-25ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 mL
  • Seq:
    2315

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light yellow to Light orange clear liquid
  • Purity (GC):
    min. 93.0 %

Specification Test Results

About this item

 :
 :100.16 g/mol
Boiling Pt:156°C
Density:0,8472 g/cm³ (20 °C)
Flash Pt:54°C
EINECS:213-190-7
UN:1987
ADR:3,III
 :GHS02
CAS Number:928-94-9
Formula:C₆H₁₂O
MDL Number:MFCD00063209