Order Entry
Finland
ContactUsLinkComponent
1-Amino-2,2,2-trifluoroethane hydrochloride ≥98.0% (by total nitrogen and titration analysis)
Irritant
1-Amino-2,2,2-trifluoroethane hydrochloride ≥98.0% (by total nitrogen and titration analysis)
Supplier:  TCI
Certificates
undefined
Irritant
1-Amino-2,2,2-trifluoroethane hydrochloride ≥98.0% (by total nitrogen and titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Light yellow to Light orange powder to crystal
  • Purity (with Total Nitrogen):
    min. 98.0 %
  • Purity (Argentmetric Titration):
    min. 98.0 %

Specification Test Results