
3,4-Dimethoxycinnamic acid ≥98.0% (by GC, titration analysis)
Catalog # TCIAD1728-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Light yellow to Green powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:181.0 to 185.0 deg-C
- Solubility in hot Methanol:very faint turbidity
Specification Test Results
About this item
:
:208.21 g/mol
Boiling Pt:343 °C (1013 hPa)
Melting Pt:198…202 °C
EINECS:219-025-5
:GHS07
CAS Number:2316-26-9
Formula:C₁₁H₁₂O₄
MDL Number:MFCD00004387