
5-Hexynoic acid ≥96.0% (by GC, titration analysis)
: TCI
:53293-00-8
:
:C₆H₈O₂
:MFCD00066346
:112.13 g/mol
:124°C
:41 °C
:1,0372 g/cm³ (20 °C)
:3265
:8,III
:GHS05
- Appearance:Colorless to Light orange to Yellow clear liquid
- Purity (GC):min. 96.0 %
- Purity (Neutralization titration):min. 96.0 %
Frequently Bought Together