4-Bromo-2-methylpyridine ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:22282-99-1
:
Formula:C₆H₆BrN
MDL Number:MFCD03788196
:172.02 g/mol
Boiling Pt:76°C
Flash Pt:79°C
:GHS07
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %