
4-Bromo-2,6-difluorobenzoic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:183065-68-1
:
Formula:C₇H₃BrF₂O₂
MDL Number:MFCD03094085
:237 g/mol
EINECS:457-940-3
:GHS07
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:200.0 to 204.0 deg-C