
4-(Trifluoromethoxy)benzaldehyde ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:659-28-9
:
Formula:C₈H₅F₃O₂
MDL Number:MFCD00041530
:190.12 g/mol
Boiling Pt:93°C
Density:1,331 g/cm³ (20 °C)
Flash Pt:70°C
EINECS:211-531-4
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 97.0 %
Specification Test Results
Frequently Bought Together