
5-Bromo-2-hydroxypyridine ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:13466-38-1
:
Formula:C₅H₄BrNO
MDL Number:MFCD00234042
:174 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:178.0 to 182.0 deg-C
Specification Test Results
Frequently Bought Together