

Ethidium bromide ≥95.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:1239-45-8
:
Formula:C₂₁H₂₀BrN₃
MDL Number:MFCD00011724
Merck Index:14,04731
Boiling Pt:72 °C (0,1 -rr)
Melting Pt:261…264 °C
Density:1,01 g/cm³ (20 °C)
Flash Pt:≤ 100 °C
UN:1544
ADR:6.1,I
:394.31 g/mol
EINECS:214-984-6
:GHS06
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Appearance:Orange to Brown to Dark purple powder to crystal
- Purity (HPLC):min. 95.0 area%
- Purity (Argentmetric Titration):min. 95.0 %
Specification Test Results
Frequently Bought Together