2-Ethylbutyric acid ≥98.0% (by GC, titration analysis)
: TCI
:88-09-5
:
:C₆H₁₂O₂
:MFCD00002670
:116.16 g/mol
:14,03110
:193°C
:–14 °C
:0,92 g/cm³ (20 °C)
:94°C
:Ambient
:201-796-4
:2810
:6.1,III
:GHS05
:GHS06
:GHS08
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %