
2-Amino-5-bromo-3-methylpyridine ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:3430-21-5
:
Formula:C₆H₇BrN₂
MDL Number:MFCD00068232
:187.04 g/mol
:GHS07
Specification Test Results
- Appearance:Light yellow to Brown powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:91.0 to 95.0 deg-C