

2,5-Difluorobenzoyl chloride ≥98.0% (by GC, titration analysis)
: TCI
:35730-09-7
:
:C₇H₃ClF₂O
:MFCD00009929
:176.55 g/mol
:93°C
:1,42 g/cm³ (20 °C)
:59°C
:252-701-8
:2920
:8,II
:GHS02
:GHS05
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 98.0 %
- Purity (Argentmetric Titration):min. 98.0 %