Order Entry
Finland
ContactUsLinkComponent
3-Chlorophenyl isocyanate ≥98.0% (by GC)
Corrosive
Toxic
Health
3-Chlorophenyl isocyanate ≥98.0% (by GC)
Catalog # TCIAI0119-25G
Supplier:  TCI
CAS Number:  
undefined
Corrosive
Toxic
Health
3-Chlorophenyl isocyanate ≥98.0% (by GC)
Catalog # TCIAI0119-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 %

Specification Test Results

About this item

 :
 :153.57 g/mol
Boiling Pt:204°C
Melting Pt:–4.4 °C
Density:1,26 g/cm³ (20 °C)
Flash Pt:82°C
Storage Temperature:Refrigerator
EINECS:220-822-5
UN:2206
ADR:6.1,II
 :GHS05
 :GHS06
 :GHS08
CAS Number:2909-38-8
Formula:C₇H₄ClNO
MDL Number:MFCD00002016