


4-Chloro-o-toluidine ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:95-69-2
:
Formula:C₇H₈ClN
MDL Number:MFCD00007842
Boiling Pt:241°C
Melting Pt:27.2 °C
Density:1,14 g/cm³ (20 °C)
EINECS:202-441-6
UN:2239
ADR:6.1,III
:141.6 g/mol
:GHS06
:GHS08
:GHS09
Specification Test Results
- Appearance:White or Colorless to Light orange to Yellow powder to lump to clear liquid
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %