
3-(Trifluoromethoxy)aniline ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:1535-73-5
:
Formula:C₇H₆F₃NO
MDL Number:MFCD00041511
:177.13 g/mol
Boiling Pt:70°C
Density:1,3254 g/cm³ (20 °C)
Flash Pt:85°C
Storage Temperature:Ambient
EINECS:216-256-3
ADR:6.1,III
:GHS07
Specification Test Results
- Appearance:White to Almost white clear liquid
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %