
4-tert-Butylthiophenol ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:2396-68-1
:
Formula:C₁₀H₁₄S
MDL Number:MFCD00022067
:166.29 g/mol
Boiling Pt:120°C
Melting Pt:–11 °C
Density:0,9917 g/cm³ (16 °C)
EINECS:219-255-6
ADR:9,
:GHS07
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 97.0 %