

(Ethyl)triphenylphosphonium bromide ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:1530-32-1
:
Formula:C₂₀H₂₀BrP
MDL Number:MFCD00011838
:371.26 g/mol
Melting Pt:205…207 °C
Storage Temperature:Ambient
EINECS:216-223-3
UN:2811
ADR:6.1,III
:GHS06
:GHS09
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:206.0 to 210.0 deg-C
- Solubility in Methanol:almost transparency