

2-Fluoro-4-methylpyridine ≥98.0% (by GC)
: TCI
:461-87-0
:
:C₆H₆FN
:MFCD00041224
:111.12 g/mol
:157°C
:1,0805 g/cm³ (25 °C)
:51°C
:1993
:3,III
:GHS02
:GHS07
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 98.0 %