
1,2-Naphthoquinone ≥95.0% (by HPLC and gravimetric)
Supplier: TCI
Certificates
About this item
CAS Number:524-42-5
:
Formula:C₁₀H₆O₂
MDL Number:MFCD00001698
:158.16 g/mol
Merck Index:14,06394
Boiling Pt:296 °C (1013 hPa)
Melting Pt:145…146 °C
Density:1,45 g/cm³ (20 °C)
EINECS:208-360-2
:GHS07
Storage Temperature:Freezer
Specification Test Results
- Appearance:Light yellow to Amber to Dark green powder to crystal
- Purity (HPLC):min. 95.0 area%
- Purity (Gravimetric Method):min. 95.0 %
- Solubility in hot Methanol:almost transparency