
2-Pyrrolidone ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:616-45-5
:
Formula:C₄H₇NO
MDL Number:MFCD00005270
:85.11 g/mol
Merck Index:14,08016
Boiling Pt:245°C
Melting Pt:24…26 °C
Density:1,1115 g/cm³ (20 °C)
Flash Pt:129 °C
EINECS:210-483-1
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White or Colorless to Almost white or Almost colorless powder to lump to clear liquid
- Purity (GC):min. 98.0 %
- Solubility in Toluene:almost transparency