Order Entry
Finland
ContactUsLinkComponent
 
 
 
2,6-Dichloro-4-(trifluoromethyl)aniline ≥98.0% (by GC)
 :  TCI
 
undefined
 
 
2,6-Dichloro-4-(trifluoromethyl)aniline ≥98.0% (by GC)
 :  TCI

 

  • Appearance:
    White to Orange to Green powder to lump
  • Purity (GC):
    min. 98.0 %