
3,5-Dimethyl-1H-pyrazole ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:67-51-6
:
Formula:C₅H₈N₂
MDL Number:MFCD00005243
:96.13 g/mol
Boiling Pt:218°C
Density:0,8839 g/cm³ (16 °C)
EINECS:200-657-5
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:106.0 to 109.0 deg-C
- Solubility in Methanol:almost transparency