
1H-Pyrazole ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:288-13-1
:
Formula:C₃H₄N₂
MDL Number:MFCD00005234
Merck Index:14,07960
Boiling Pt:186 °C (1013 hPa)
Melting Pt:66…68 °C
Density:1,34 g/cm³ (20 °C)
:68.08 g/mol
EINECS:206-017-1
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:67.0 to 71.0 deg-C
- Solubility in Water:almost transparency
Specification Test Results
Frequently Bought Together