
1-Methylisoquinoline ≥98.0% (by GC, titration analysis)
: TCI
:1721-93-3
:
:C₁₀H₉N
:MFCD00006902
:143.19 g/mol
:255°C
:10 °C
:1,078 g/cm³ (25 °C)
:102°C
:217-017-6
:GHS07
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 98.0 %
Frequently Bought Together