


Glycolic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:79-14-1
:
Formula:C₂H₄O₃
MDL Number:MFCD00004312
:76.05 g/mol
Merck Index:14,04498
Boiling Pt:266 °C (1013 hPa)
Melting Pt:79…80 °C
Density:1,49 g/cm³ (20 °C)
Flash Pt:≥ 300 °C
Storage Temperature:Ambient
EINECS:201-180-5
UN:3261
ADR:8,II
:GHS05
:GHS07
:GHS08
Specification Test Results
- Appearance:White powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:75.0 to 80.0 deg-C
- Solubility in Water:almost transparency